WHEATON® Bottles, Wide Neck, with Screw Cap
Amber, USP Type III soda-lime glass.
- Easy removal of contents
- White polypropylene screw cap
- Hazardous communication label included with each case of bottles allows for content ID and safety notification
- Tamper-evident seal included with each case of bottles keeps bottle sealed which protecting contents from accidental misuse
Ideal for storage of light-sensitive samples or dry powders.
With poly-vinyl lined PP cap
- CapacityThreadExt.ØHeightPk
- 215-4517W21693930 ml28-40037 mm65 mm241 * 24 Itemskr 970,00: W216939: WHEAW216939
Specifications
Environmentally Preferable - 215-4518W21694060 ml33-40044 mm75 mm241 * 24 Itemskr 950,00: W216940: WHEAW216940
Specifications
Environmentally Preferable - 215-4519W216941125 ml38-40054 mm95 mm241 * 24 Itemskr 1 100,00: W216941: WHEAW216941
Specifications
Environmentally Preferable - 215-4520W216942250 ml45-40066 mm119 mm241 * 24 Itemskr 1 350,00: W216942: WHEAW216942
Specifications
Environmentally Preferable - 215-4521W216943500 ml53-40080 mm146 mm121 * 12 Itemskr 910,00: W216943: WHEAW216943
Specifications
Environmentally Preferable - 215-4522W2169441000 ml53-40099 mm178 mm121 * 12 Itemskr 1 275,00: W216944: WHEAW216944
Specifications
Environmentally Preferable
With poly seal lined phenolic cap
- CapacityThreadExt.ØHeightPk
- 612-6855W21695260 ml33-40044 mm75 mm241 * 24 Itemskr 1 050,00: W216952: WHEAW216952
Specifications
Environmentally Preferable - 612-6856W216953125 ml38-40054 mm95 mm241 * 24 Itemskr 1 375,00: W216953: WHEAW216953
Specifications
Environmentally Preferable - 612-6854W21695130 ml28-40037 mm65 mm241 * 24 Itemskr 1 125,00: W216951: WHEAW216951
Specifications
Environmentally Preferable
With Teflon® faced foamed PE lined PP cap
- CapacityThreadExt.ØHeightPk
- WHEAW216949W216949500 ml53-40080 mm146 mm121 * 12 Itemskr 1 375,00: W216949: WHEAW216949
Specifications
Environmentally Preferable - W2169501000 ml53-40099 mm178 mm121 * 12 Itemskr 1 700,00: W216950: WHEAW216950
Specifications
Environmentally Preferable - W21694530 ml28-40037 mm65 mm241 * 24 Itemskr 1 275,00: W216945: WHEAW216945
Specifications
Environmentally Preferable - 215-4135W21694660 ml33-40044 mm75 mm241 * 24 Itemskr 1 300,00: W216946: WHEAW216946
Specifications
Environmentally Preferable - 612-5881W216947125 ml38-40054 mm95 mm241 * 24 Itemskr 1 550,00: W216947: WHEAW216947
Specifications
Environmentally Preferable - W216948250 ml45-40066 mm119 mm241 * 24 Itemskr 1 950,00: W216948: WHEAW216948
Specifications
Environmentally Preferable
Without cap
- CapacityThreadExt.ØHeightPk
- WHEAW216962W216962125 ml38-40054 mm95 mm1801 * 180 Itemskr 5 050,00: W216962: WHEAW216962
Specifications
Environmentally Preferable - WHEAW216960W21696030 ml28-40037 mm65 mm4321 * 432 Itemskr 9 575,00: W216960: WHEAW216960
Specifications
Environmentally Preferable - : W216961: WHEAW216961
Specifications
Environmentally Preferable