Dropping bottles, LDPE, Wheaton®
Supplier: DWK Life Sciences
Certificates
Dropping bottles made of natural or white LDPE. The white LDPE bottles tends to protect the sensitive samples from UV light.
- Good chemical resistance
- Good temperature resistance
Natural
Product Information
- Catalog #Supplier #CapacityHeightExt.ØThreadPkAvailabilityPrice
- WHEAW242821-AW242821-A3 ml39 mm14 mm8-4251.0001 * 1.000 Itemskr 25 825,00Supplier Article Number: W242821-ALocal Article Number: WHEAW242821-A
Specifications
Environmentally Preferable - 215-4137W2428213 ml39 mm14 mm8-4251001 * 100 Itemskr 3 275,00Supplier Article Number: W242821Local Article Number: WHEAW242821
Specifications
Environmentally Preferable - WHEAW242822-AW242822-A6 ml42 mm18 mm13-4251.0001 * 1.000 Itemskr 26 075,00Supplier Article Number: W242822-ALocal Article Number: WHEAW242822-A
Specifications
Environmentally Preferable - WHEAW242823-AW242823-A7 ml51 mm19 mm15-4151.0001 * 1.000 Itemskr 27 325,00Supplier Article Number: W242823-ALocal Article Number: WHEAW242823-A
Specifications
Environmentally Preferable - WHEAW242825-AW242825-A15 ml66 mm25 mm15-4151.0001 * 1.000 Itemskr 31 125,00Supplier Article Number: W242825-ALocal Article Number: WHEAW242825-A
Specifications
Environmentally Preferable - 215-4139W2428226 ml42 mm18 mm13-4251001 * 100 Itemskr 3 425,00Supplier Article Number: W242822Local Article Number: WHEAW242822
Specifications
Environmentally Preferable - 215-4141W2428237 ml51 mm19 mm15-4151001 * 100 Itemskr 3 450,00Supplier Article Number: W242823Local Article Number: WHEAW242823
Specifications
Environmentally Preferable - 215-4143W24282410 ml46 mm25 mm15-4151001 * 100 Itemskr 3 600,00Supplier Article Number: W242824Local Article Number: WHEAW242824
Specifications
Environmentally Preferable - WHEAW242824-AW242824-A10 ml46 mm25 mm15-4151.0001 * 1.000 Itemskr 28 850,00Supplier Article Number: W242824-ALocal Article Number: WHEAW242824-A
Specifications
Environmentally Preferable - 215-4145W24282515 ml66 mm25 mm15-4151001 * 100 Itemskr 3 775,00Supplier Article Number: W242825Local Article Number: WHEAW242825
Specifications
Environmentally Preferable - 215-4147W24282630 ml70 mm32 mm20-4101001 * 100 Itemskr 4 500,00Supplier Article Number: W242826Local Article Number: WHEAW242826
Specifications
Environmentally Preferable - WHEAW242826-AW242826-A30 ml70 mm32 mm20-4101.0001 * 1.000 Itemskr 40 475,00Supplier Article Number: W242826-ALocal Article Number: WHEAW242826-A
Specifications
Environmentally Preferable - 215-4149W24282760 ml85 mm39 mm20-4101001 * 100 Itemskr 5 125,00Supplier Article Number: W242827Local Article Number: WHEAW242827
Specifications
Environmentally Preferable
White
Product Information
- Catalog #Supplier #CapacityHeightExt.ØThreadPkAvailabilityPrice
- 215-4136W2428313 ml39 mm14 mm8-4251001 * 100 Itemskr 3 275,00Supplier Article Number: W242831Local Article Number: WHEAW242831
Specifications
Environmentally Preferable - 215-4140W2428337 ml51 mm19 mm15-4151001 * 100 Itemskr 3 450,00Supplier Article Number: W242833Local Article Number: WHEAW242833
Specifications
Environmentally Preferable - WHEAW242833-AW242833-A7 ml51 mm19 mm15-4151.0001 * 1.000 Itemskr 27 325,00Supplier Article Number: W242833-ALocal Article Number: WHEAW242833-A
Specifications
Environmentally Preferable - WHEAW242831-AW242831-A3 ml39 mm14 mm8-4251.0001 * 1.000 Itemskr 25 825,00Supplier Article Number: W242831-ALocal Article Number: WHEAW242831-A
Specifications
Environmentally Preferable - 215-4142W24283410 ml46 mm25 mm15-4151001 * 100 Itemskr 3 600,00Supplier Article Number: W242834Local Article Number: WHEAW242834
Specifications
Environmentally Preferable - WHEAW242834-AW242834-A10 ml46 mm25 mm15-4151.0001 * 1.000 Itemskr 28 850,00Supplier Article Number: W242834-ALocal Article Number: WHEAW242834-A
Specifications
Environmentally Preferable - 215-4138W2428326 ml42 mm18 mm13-4251001 * 100 Itemskr 3 425,00Supplier Article Number: W242832Local Article Number: WHEAW242832
Specifications
Environmentally Preferable - WHEAW242832-AW242832-A6 ml42 mm18 mm13-4251.0001 * 1.000 Itemskr 26 075,00Supplier Article Number: W242832-ALocal Article Number: WHEAW242832-A
Specifications
Environmentally Preferable - 215-4144W24283515 ml66 mm25 mm15-4151001 * 100 Itemskr 3 875,00Supplier Article Number: W242835Local Article Number: WHEAW242835
Specifications
Environmentally Preferable - WHEAW242835-AW242835-A15 ml66 mm25 mm15-4151.0001 * 1.000 Itemskr 31 125,00Supplier Article Number: W242835-ALocal Article Number: WHEAW242835-A
Specifications
Environmentally Preferable - 215-4146W24283630 ml70 mm32 mm20-4101001 * 100 Itemskr 4 500,00Supplier Article Number: W242836Local Article Number: WHEAW242836
Specifications
Environmentally Preferable - WHEAW242836-AW242836-A30 ml70 mm32 mm20-4101.0001 * 1.000 Itemskr 40 475,00Supplier Article Number: W242836-ALocal Article Number: WHEAW242836-A
Specifications
Environmentally Preferable - 215-4148W24283760 ml85 mm39 mm20-4101001 * 100 Itemskr 5 125,00Supplier Article Number: W242837Local Article Number: WHEAW242837
Specifications
Environmentally Preferable