Surgical masks, Profile™ PFM
Leverandør: Ansell-Nitritex
NITRPFML-2050EA 1350 NOK
NITRPFML-2050
Surgical masks, Profile™ PFM
Profile™ PFM surgical face mask with highly efficient bacterial filtration.
  • Hydrophobic outer layer (175 mm)
  • Latex-free hydrophilic inner layer
  • Three layer construction