Infrared thermometer, Traceable®
620-2096EA 4325 NOK
VWRI620-2096
Infrared thermometer, Traceable®
This infrared thermometer provides the widest possible range for accurately measuring different surface temperatures.
  • Field of view of 7:1
  • Adjustable emissivity from 0,2 to 1,0 in increments of 0,01
  • Data HOLD, maximum/minimum memory, and one-second sampling
Infrared sensor and type-K beaded probe.
Leveringsinformasjon: Supplied with a Traceable® certificate, K-type beaded probe, carrying case, and nine-volt battery.