Digital thermo-hygrometer, Traceable® Jumbo
628-0031EA 1075 NOK
VWRI628-0031
Digital thermo-hygrometer, Traceable® Jumbo
This unit continuously and simultaneously displays ambient relative humidity and temperatures. The changes are shown in less than 30 seconds, for monitoring conditions overnight, on weekends, or any other time period.
  • Easy to read jumbo display (25 mm)
  • Probe and cable perform accurately under water
  • Minimum/maximum memories displayed or cleared at any time with the touch of a button
Internal sensor and external probe with cable (3 metres).
Sertifiseringer: Compliant with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
Leveringsinformasjon: Supplied with flip-open stand, wall mount, AAA batteries and Traceable® certificate of calibration.