Norway
Order Entry
Industrial filter boards, Munktell
Industrial filter boards, Munktell
Supplier:  Ahlstrom-Munksjö
Certificates
undefined
Industrial filter boards, Munktell
Supplier:  Ahlstrom-Munksjö
Specifications
  • Catalog No:
  • 512-6202
  • 512-6200
  • BINZ3.10389.025
  • Grade:
  • 1220
  • 1220
  • 1220
  • Weight:
  • 475 g/m²
  • 475 g/m²
  • 475 g/m²
  • Thickness:
  • 1,10 µm
  • 1,10 µm
  • 1,10 µm
  • Capillary rise:
  • 120 mm/10 min
  • 120 mm/10 min
  • 120 mm/10 min
Specifications
Grade 1220
Product Information
  • Catalog #
    Supplier #
    Grade
    Size
    Pk
    Availability
    Price
  • 512-6202
    231012
    1220
    430×430 mm
    50
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - kr 370,00
    kr 370,00
    1 * 50 Items
       kr 370,00
    Specifications:
    More
    Specifications
    Grade
    1220
    Environmentally Preferable
    Weight
    475 g/m²
    Thickness
    1,10 µm
    Capillary rise
    120 mm/10 min
  • 512-6200
    231001
    1220
    25 mm
    100
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - kr 340,00
    kr 340,00
    1 * 100 Items
       kr 340,00
    Specifications:
    More
    Specifications
    Grade
    1220
    Environmentally Preferable
    Weight
    475 g/m²
    Thickness
    1,10 µm
    Capillary rise
    120 mm/10 min
  • BINZ3.10389.025
    3.10389.025
    1220
    25 mm
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 Items
    1 * 1.000 Items - kr 5 125,00
    kr 5 125,00
    1 * 1.000 Items
       kr 5 125,00
    Specifications:
    More
    Specifications
    Grade
    1220
    Environmentally Preferable
    Weight
    475 g/m²
    Thickness
    1,10 µm
    Capillary rise
    120 mm/10 min