/
Norway
Products
Services
Digital Solutions
Applications
Support
About
kr 0,00
Order Entry
Disposable earplugs, uvex x-fit detec
UVEX2112.011
:
UVEX
Disposable earplugs, uvex x-fit detec
UVEX2112.011
:
UVEX
kr 2 050,00
1 * 100 PAR
-
+
Colour:
Blue
Description:
Detectable earplugs with cord
Pk:
100 PAIR