Tubing, PTFE, heat shrinkable, BOLA
PTFE, translucent.
- Temperature resistance: from −200 to +250 °C
- Universal chemical resistance
- Shrink rate: 4:1
- Incombustible (ASTM D876)
- Insulation resistance 1018 O/cm (ASTM D 876)
- Good electric strength
- Flame retardant
For protection of probes, cables, electric components etc. against chemical disturbance. The shrink rate of 4:1 means that the inner diameter of the tubing shrinks to approx. ¼ of the original inner diameter and that the length shrinks to approx. 15%. Good heat transmission due to low wall thickness.
- Tubing int.ØTubing materialPk
- Heat-shrinkable tubingHeat-shrinkable tubingBOHLS1828-56S1828-5619,00 mmPTFE1 m1 * 1 mkr 975,00
Specifications
Environmentally Preferable - Heat-shrinkable tubingHeat-shrinkable tubingBOHLS1828-64S1828-6425,40 mmPTFE1 m1 * 1 mkr 1 075,00
Specifications
Environmentally Preferable - Heat-shrinkable tubingHeat-shrinkable tubingBOHLS1828-08S1828-082,00 mmPTFE1 m1 * 1 mkr 520,00
Specifications
Environmentally Preferable - Heat-shrinkable tubingHeat-shrinkable tubingBOHLS1828-24S1828-244,70 mmPTFE1 m1 * 1 mkr 540,00
Specifications
Environmentally Preferable - Heat-shrinkable tubingHeat-shrinkable tubingBOHLS1828-16S1828-163,20 mmPTFE1 m1 * 1 mkr 635,00
Specifications
Environmentally Preferable - Heat-shrinkable tubingHeat-shrinkable tubingBOHLS1828-40S1828-409,50 mmPTFE1 m1 * 1 mkr 685,00
Specifications
Environmentally Preferable - Heat-shrinkable tubingHeat-shrinkable tubingBOHLS1828-48S1828-4812,70 mmPTFE1 m1 * 1 mkr 810,00
Specifications
Environmentally Preferable - Heat-shrinkable tubingHeat-shrinkable tubingBOHLS1828-32S1828-326,30 mmPTFE1 m1 * 1 mkr 505,00
Specifications
Environmentally Preferable