Rubber protection mats
Black protection sheets made from fluoroelastomer rubber (FPM). Durometer 75 Shore A.
- For temperature ranges from −10 to over +200 °C
- Good resistance to ozone and weathering
- Benzine and mineral oil resistant
- ColourL×WThicknessPk
- DENE52020015202001Black200×200 mm1,00 mm11 * 1 Itemskr 1 600,00: 5202001: DENE5202001
Specifications
Environmentally Preferable - DENE52020155202015Black200×200 mm1,50 mm11 * 1 Itemskr 2 225,00: 5202015: DENE5202015
Specifications
Environmentally Preferable - DENE52020025202002Black200×200 mm2,00 mm11 * 1 Itemskr 2 550,00: 5202002: DENE5202002
Specifications
Environmentally Preferable - DENE52020035202003Black200×200 mm3,00 mm11 * 1 Itemskr 2 900,00: 5202003: DENE5202003
Specifications
Environmentally Preferable - DENE52020045202004Black200×200 mm4,00 mm11 * 1 Itemskr 4 825,00: 5202004: DENE5202004
Specifications
Environmentally Preferable - DENE52030015203001Black300×300 mm1,00 mm11 * 1 Itemskr 2 925,00: 5203001: DENE5203001
Specifications
Environmentally Preferable - DENE52030155203015Black300×300 mm1,50 mm11 * 1 Itemskr 2 950,00: 5203015: DENE5203015
Specifications
Environmentally Preferable - DENE52030025203002Black300×300 mm2,00 mm11 * 1 Itemskr 3 775,00: 5203002: DENE5203002
Specifications
Environmentally Preferable - DENE52030035203003Black300×300 mm3,00 mm11 * 1 Itemskr 4 800,00: 5203003: DENE5203003
Specifications
Environmentally Preferable - DENE52030045203004Black300×300 mm4,00 mm11 * 1 Itemskr 6 450,00: 5203004: DENE5203004
Specifications
Environmentally Preferable
This product has been blocked by your organization. Please contact your purchasing department for more information