Stirrer blades, SP Bel-Art
Leverandør: Bel-Art Products, a part of SP
SP Bel-Art
BELAF379720000EA 315 NOK
BELAF379720000
Stirrer blades, SP Bel-Art
Omrørere Omrørerblad
Stirrer blade made of PTFE. Fits all standard button-tip stirring shafts.
Long groove keyhole provides easy insertion into flask neck and then rotates into proper position for stirring action.