Top loader oven, round, Top 16/R
Leverandør: Nabertherm
NABETOP16/REA 0 NOK
NABETOP16/R
Top loader oven, round, Top 16/R
The small but powerful Top 16/R is the ideal kiln for hobby ceramics, porcelain painting, and for small fusing jobs.
  • B400 controller for easy and individual programming
  • Heating elements embedded in grooves, heating from all sides
  • Rapid switching cycles result in precise temperature control
  • Lightweight refractory bricks inside furnace chamber for clean firing results
  • Attractive design with low-maintenance stainless steel housing
  • Lid with adjustable quick-release lock and padlock hasp
Also suitable for glazing samples and small pieces