Robotspiss for FLIPR Liberty 384 (type B)
Leverandør: Biotix
732-2414EA 7275 NOK
BITXF-0030-3SB
Robotspiss for FLIPR Liberty 384 (type B)
Biotix robotic tips for Molecular Devices® automated platforms are manufactured to the highest engineering standards for robotic systems. The ultra-straight tips target the centre of microplate wells precisely, delivering exceptional accuracy and precision without risk of jamming.
  • Transparent or black PP, sterile
  • High clarity X-Resin™ has superior low retention performance
  • Blade™ technology eliminates hanging drops at the distal end of the pipette tip
  • Each lot is tested in Biotix's in-house automation laboratory
  • Tips are BioReady™ certified to be free of RNase, DNase, and Endotoxins (pyrogens)
With specifications to ensure complete compatibility, no changes in programming scripts should be necessary to use Biotix tips; use of the standard definitions bundled in the robotic software program is appropriate.
These tips are compatible with Molecular Devices® FLIPR® Liberty 384 Head.