Buffer, TRIS-glycine, VWR
0251-2PKEA 745 NOK
VWRC0251-2PK
Buffer, TRIS-glycine, VWR
A pre-blended Tris-glycine powder for preparation of Laemmli SDS-PAGE running buffer.
The 40 L pack size prepares 40 litres of 1X Tris-glycine solution (0,025 M Tris base and 0,192 M glycine). Each Ready-Pack™ prepares 1 litre of a 10X concentrate.