Skip to Header
Skip to Main Content
Skip to Footer
/
Norway
Products
Services
Digital Solutions
Applications
Support
About
kr 0,00
Order Entry
Accessories for Nova Safety
614-0173
:
GERBER FUNKE
Accessories for Nova Safety
614-0173
:
GERBER FUNKE
: 3641
: GERB3641
kr 225,00
1 * 1 Items
-
+
Description:
Replacement tube for butyrometer, brass, with flanged edge, ext./int. Ø×L: 27/25,8×170 mm
Pk:
1