Anemometers, testo 512
Leverandør: Testo
TEST0560.5126EA 6975 NOK
TEST0560.5126 TEST0560.5127 TEST0560.5128 620-0199
Anemometers, testo 512
Anemometere
Easy-to-read, large and backlit display.
Measurement data is printed on site with date and time along with minimum/maximum values.
A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
They are used to display pressure and flow velocity.
Sertifiseringer: NIST traceable certification.