Sleeve protectors, cut resistant, Aracut®
Leverandør: Honeywell Safety
Aracut® Sleeves
DALL4402835EA 500 NOK
DALL4402835
Sleeve protectors, cut resistant, Aracut®
Arbeidstøy
Knitted, double Kevlar® sleeve with thumb hole, to protect the underarm and palm against cuts.
  • Close-fitting sleeve with good tactile sensitivity, optimum wearer comfort and increased strength
  • Kevlar® fibre offers excellent cut resistance and protects against thermal risks (contact heat)
  • Double thickness increases the mechanical and thermal resistance
  • Length guarantees full protection of the forearm
Ideal for use in the glass, iron and steel industry, in metalworking, in the paper and wood industry, in shipbuilding and in the construction industry.
Gauge: 10
Sertifiseringer: EN 407 (X1XXXX), EN 388 (132X), Cat. II