Nitrogen receivers (Schlenk flasks), with PTFE needle-valve stopcock
Leverandør: Lenz
DURAN® borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With conical ground joint, PTFE needle-valve stopcock and inscription label.
BESTILL
Utvid alle / Skjul alle
Kapasitet Hylsestr Fp Art.nr Antall
25 ml 14/23 NS 1 LENZ03360622
50 ml 14/23 NS 1 LENZ03360628
50 ml 29/32 NS 1 LENZ03360828
100 ml 29/32 NS 1 LENZ03360837
250 ml 29/32 NS 1 LENZ03360849
500 ml 29/32 NS 1 LENZ03360858
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall