Film dressing, Tegaderm™
Leverandør: 3M Medical
MMMEM16004EA 2125 NOK
MMMEM16004
Film dressing, Tegaderm™
This non sterile transparent film dressing offers all the benefits of surgical tape while providing a waterproof, bacterial, and viral barrier.
  • Provides a waterproof barrier to external contaminants including liquids, bacteria, and viruses
  • Can be cut and customised to desired length
  • Can be used for fixation of medical devices or infusion tubing
  • Conformable design fits body contours and permits free movement without stress on the skin
Suitable applications include: Fixation of medical devices (e.g. drainage tubes, etc.), waterproof fixation of primary dressings such as gauzes, alginates, foams, etc. and protection of at-risk skin from friction and body fluids.
Please note: the dressing is not recommended as a primary wound or IV dressing.