Air sampling device
Leverandør: Gastec
GASTGSP-300FT-2EA 35250 NOK
GASTGSP-300FT-2
Air sampling device
Prøvetakere Luftprøvetakere
Small and compact air sampling pump with an integrated flowmetre for hand-held, personal or on-site sampling.
  • Constant flow rate for 10 hours continuous sampling.
  • Extremely low noise air pump and automatic shut-off air pump
  • Integrated flow rate and accumulated time