/
Norway
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Chromium standard solution, 1,000 mg/L Cr in dil. nitric acid (from Cr(NO₃)₃.9H₂O) standard for AAS and ICP
PERCN9300112
:
PerkinElmer
:
Chromium standard solution, 1,000 mg/L Cr in dil. nitric acid (from Cr(NO₃)₃.9H₂O) standard for AAS and ICP
PERCN9300112
:
PerkinElmer
:
kr 2 525,00
1 * 1 Items
-
+
Concentration:
1000 mg/L
Matrix:
2% HNO₃
Pk:
1
Element:
Chromium
Symbol:
Cr