Plantevekstskap, KBW-serien
Leverandør: Binder
BIND9020-0339EA 0 NOK
BIND9020-0339 BIND9020-0338 BIND9020-0340
Plantevekstskap, KBW-serien
Testkammer Plantevekstkammer
The KBW series plant growth chambers are precise units for plant cultivation under advanced working conditions. They provide perfect simulation of a wide range of growing environments. The units satisfy all requirements for optimum lighting and temperature conditions, so that culture processes can be defined and reproduced exactly. The design includes two or three variable position daylight cassettes with five daylight luminescence tubes each, which provides homogeneous light distribution. Units self-test for comprehensive status analysis.
  • Precision and outstanding dynamics
  • Even distribution of light on all levels and adjustable fan speed
  • Environmentally friendly
  • Intuitive touchscreen controller with time-segment and real-time programming
  • Ethernet interface for APT-COM™ DataControlSystem communication software
The KBW models have a heated inner glass door, an access port with silicone plug Ø30 mm on the left hand side. Units feature an independent adjustable temperature safety device Class 3.1 with visual and audible alarm, which protects the samples and the incubator. Microprocessor controller with two programs each with 10 sections or, alternatively, switch over to one program with 20 sections. Digital temperature setting with an accuracy of a tenth of a degree, integrated weekly program timer, elapsed time indicator and electronically controlled APT.line™ preheating chamber ensures temperature accuracy and reproducible results.
Values measured via the internal data logger can be read out in open format via USB. All models have an tight sealing inner door made of ESG safety glass, the door is heated against condensation. Glass corrosion is avoided thanks to the special TIMELESS coating. The units have 4 casters each, 2 with brakes them.
Leveringsinformasjon: Supplied with two (or three) stainless steel shelves and Binder test certificate. Calibration and validation is possible, for details please contact your local Avantor sales office.