Chrome security truck, Super Erecta®
- ForW×D×HPk
- Mobile security truck Super Erecta®Mobile security truck Super Erecta®139-0804SEC53DC914×610 mm shelves1035×692×1726 mm11 * 1 Itemskr 29 200,00
Specifications
Environmentally Preferable - Mobile security truck Super Erecta®Mobile security truck Super Erecta®139-0366SEC53EC914×610 mm shelves1035×692×1726 mm11 * 1 Itemskr 29 425,00
Specifications
Environmentally Preferable - Mobile security truck Super Erecta®Mobile security truck Super Erecta®139-0367SEC55EC1219×610 mm shelves1340×692×1726 mm11 * 1 Itemskr 35 600,00
Specifications
Environmentally Preferable - Mobile security truck Super Erecta®Mobile security truck Super Erecta®139-0023SEC56DC1524×610 mm shelves1651×692×1726 mm11 * 1 Itemskr 37 775,00
Specifications
Environmentally Preferable - Mobile security truck Super Erecta®Mobile security truck Super Erecta®139-0368SEC56EC1524×610 mm shelves1651×692×1726 mm11 * 1 Itemskr 38 075,00
Specifications
Environmentally Preferable - Mobile security truck Super Erecta®Mobile security truck Super Erecta®METOSEC66ECSEC66EC1524×760 mm shelves1651×851×1740 mm11 * 1 Itemskr 47 625,00
Specifications
Environmentally Preferable