VWR®, Funnels with Short Stem, Disposable
PP or PS.
- Good chemical resistance
- Good temperature resistance
- PP funnels are autoclavable
Suitable for use with standard filter paper.
PP
Product Information
- Catalog #MaterialLengthStem lengthStem O-ØFunnel O-ØPkAvailabilityPrice
- 221-025174,4 mm22,7 mm7,4/4,3 mm65 mm1001 * 100 Itemskr 860,00
Specifications
Environmentally Preferable - 221-025271,3 mm27,1 mm7,4/4,3 mm55 mm1001 * 100 Itemskr 860,00
Specifications
Environmentally Preferable
PS
Product Information
- Catalog #MaterialLengthStem lengthStem O-ØFunnel O-ØPkAvailabilityPrice
- 221-0253Polystyrene71,3 mm27,1 mm7,4/4,3 mm55 mm1001 * 100 Itemskr 625,00
Specifications
Environmentally Preferable