3-Pyridylacetonitrile
Leverandør: Molekula
MOLE52960883-25GEA
0
NOK
MOLE52960883-25G
3-Pyridylacetonitrile
3-Pyridylacetonitrile
Formel:
C₇H₆N₂ MW: 118,14 g/mol |
MDL nummer:
MFCD00006406 CAS-nummer: 6443-85-2 EINECS: 229-241-1 FN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...