Norway
Order Entry
Urine collection tubes, vacuum, Vacutest®
undefined
Urine collection tubes, vacuum, Vacutest®
Tubes without urine preservation, conical
  •  
    Capacity
    Version
    Ext.Ø
    Height
    Pk
  • 216-1799
    14850
    9,5 ml
    Yellow cap
    16 mm
    100 mm
    100
    1 * 100 Items
    1 * 100 Items - kr 505,00
    kr 505,00
    1 * 100 Items
       kr 505,00
     : 14850
     : KIMA14850
    Specifications
    Environmentally Preferable
  • KIMA148730
    148730
    9,5 ml
    red cap
    16 mm
    100 mm
    100
    1 * 100 Items
    1 * 100 Items - kr 450,00
    kr 450,00
    1 * 100 Items
       kr 450,00
     : 148730
     : KIMA148730
    Specifications
    Environmentally Preferable
  • KIMA148710
    148710
    9,5 ml
    White cap
    16 mm
    100 mm
    100
    1 * 100 Items
    1 * 100 Items - kr 450,00
    kr 450,00
    1 * 100 Items
       kr 450,00
     : 148710
     : KIMA148710
    Specifications
    Environmentally Preferable
Tubes without urine preservative, cylindrical
  •  
    Capacity
    Version
    Ext.Ø
    Height
    Pk
  • 216-1483
    14930
    9,0 ml
    Yellow cap
    16 mm
    100 mm
    100
    1 * 100 Items
    1 * 100 Items - kr 415,00
    kr 415,00
    1 * 100 Items
       kr 415,00
     : 14930
     : KIMA14930
    Specifications
    Environmentally Preferable
  • KIMA14905
    14905
    4,0 ml
    Red cap
    13 mm
    75 mm
    100
    1 * 100 Items
    1 * 100 Items - kr 295,00
    kr 295,00
    1 * 100 Items
       kr 295,00
     : 14905
     : KIMA14905
    Specifications
    Environmentally Preferable
  • 216-1801
    14910
    6,0 ml
    Yellow cap
    13 mm
    100 mm
    100
    1 * 100 Items
    1 * 100 Items - kr 355,00
    kr 355,00
    1 * 100 Items
       kr 355,00
     : 14910
     : KIMA14910
    Specifications
    Environmentally Preferable
  • 216-1804
    148201
    6,0 ml
    Yellow cap
    16 mm
    100 mm
    100
    1 * 100 Items
    1 * 100 Items - kr 415,00
    kr 415,00
    1 * 100 Items
       kr 415,00
     : 148201
     : KIMA148201
    Specifications
    Environmentally Preferable
  • KIMA14922D
    14922D
    6,0 ml
    Red cap
    13 mm
    100 mm
    100
    1 * 100 Items
    1 * 100 Items - kr 340,00
    kr 340,00
    1 * 100 Items
       kr 340,00
     : 14922D
     : KIMA14922D
    Specifications
    Environmentally Preferable
  • KIMA14936
    14936
    9,0 ml
    Beige cap
    16 mm
    100 mm
    100
    1 * 100 Items
    1 * 100 Items - kr 415,00
    kr 415,00
    1 * 100 Items
       kr 415,00
     : 14936
     : KIMA14936
    Specifications
    Environmentally Preferable
  • 216-1795
    14900
    4,0 ml
    Yellow cap
    13 mm
    75 mm
    100
    1 * 100 Items
    1 * 100 Items - kr 355,00
    kr 355,00
    1 * 100 Items
       kr 355,00
     : 14900
     : KIMA14900
    Specifications
    Environmentally Preferable
  • KIMA149400
    149400
    9,0 ml
    Red cap
    16 mm
    100 mm
    100
    1 * 100 Items
    1 * 100 Items - kr 415,00
    kr 415,00
    1 * 100 Items
       kr 415,00
     : 149400
     : KIMA149400
    Specifications
    Environmentally Preferable
Tubes with urine preservative
  •  
    Capacity
    Version
    Ext.Ø
    Height
    Pk
  • 216-1803
    149610
    4 ml
    Yellow cap
    13 mm
    75 mm
    100
    1 * 100 Items
    1 * 100 Items - kr 390,00
    kr 390,00
    1 * 100 Items
       kr 390,00
     : 149610
     : KIMA149610
    Specifications
    Environmentally Preferable
  • 216-1802
    149600
    9,0 ml
    Yellow cap
    16 mm
    100 mm
    100
    1 * 100 Items
    1 * 100 Items - kr 485,00
    kr 485,00
    1 * 100 Items
       kr 485,00
     : 149600
     : KIMA149600
    Specifications
    Environmentally Preferable
  • 216-1800
    148600
    9,0 ml
    Yellow cap
    16 mm
    100 mm
    100
    1 * 100 Items
    1 * 100 Items - kr 550,00
    kr 550,00
    1 * 100 Items
       kr 550,00
     : 148600
     : KIMA148600
    Specifications
    Environmentally Preferable