Beakers, low or tall form
KAVA632417012940
: Kavalier
: 632417012940
Low form
- CapacityHeightExt.ØPk
- 213-1041N6324271500055 ml30 mm22 mm101 * 10 Itemskr 1 025,00: N632427150005: KAVAN632427150005
Specifications
Environmentally Preferable - 213-1042N63242715001010 ml35 mm26 mm101 * 10 Itemskr 1 025,00: N632427150010: KAVAN632427150010
Specifications
Environmentally Preferable - 213-1043N63242715002525 ml50 mm34 mm101 * 10 Itemskr 835,00: N632427150025: KAVAN632427150025
Specifications
Environmentally Preferable - 213-1046N632427150150150 ml80 mm60 mm101 * 10 Itemskr 1 050,00: N632427150150: KAVAN632427150150
Specifications
Environmentally Preferable - 213-1047632417010250250 ml95 mm70 mm101 * 10 Itemskr 955,00: 632417010250: KAVA632417010250
Specifications
Environmentally Preferable - 213-1048632417010400400 ml110 mm80 mm101 * 10 Itemskr 1 075,00: 632417010400: KAVA632417010400
Specifications
Environmentally Preferable - 213-1049632417010600600 ml125 mm90 mm101 * 10 Itemskr 1 300,00: 632417010600: KAVA632417010600
Specifications
Environmentally Preferable - 213-10516324170109401000 ml145 mm105 mm101 * 10 Itemskr 2 100,00: 632417010940: KAVA632417010940
Specifications
Environmentally Preferable - KAVA6324170109506324170109502000 ml185 mm130 mm41 * 4 Itemskr 1 425,00: 632417010950: KAVA632417010950
Specifications
Environmentally Preferable - KAVA6324170109566324170109565000 ml270 mm170 mm21 * 2 Itemskr 4 150,00: 632417010956: KAVA632417010956
Specifications
Environmentally Preferable - KAVA63241101096663241101096610000 ml350 mm217 mm11 * 1 Itemskr 3 725,00: 632411010966: KAVA632411010966
Specifications
Environmentally Preferable - 213-1050632417010800800 ml135 mm100 mm101 * 10 Itemskr 1 800,00: 632417010800: KAVA632417010800
Specifications
Environmentally Preferable - KAVA6324170109526324170109523000 ml210 mm150 mm41 * 4 Itemskr 6 200,00: 632417010952: KAVA632417010952
Specifications
Environmentally Preferable
Tall form
- CapacityHeightExt.ØPk
- 213-1063N632427152150150 ml95 mm54 mm101 * 10 Itemskr 1 100,00: N632427152150: KAVAN632427152150
Specifications
Environmentally Preferable - KAVA6324170129406324170129401000 ml180 mm95 mm101 * 10 Itemskr 1 725,00: 632417012940: KAVA632417012940
Specifications
Environmentally Preferable - KAVA6324170129506324170129502000 ml240 mm120 mm61 * 6 Itemskr 1 850,00: 632417012950: KAVA632417012950
Specifications
Environmentally Preferable - 213-1062N632427152100100 ml80 mm48 mm101 * 10 Itemskr 835,00: N632427152100: KAVAN632427152100
Specifications
Environmentally Preferable - KAVA632417012250632417012250250 ml120 mm60 mm101 * 10 Itemskr 830,00: 632417012250: KAVA632417012250
Specifications
Environmentally Preferable - 213-1065632417012400400 ml130 mm70 mm101 * 10 Itemskr 1 125,00: 632417012400: KAVA632417012400
Specifications
Environmentally Preferable - 213-1066632417012600600 ml150 mm80 mm101 * 10 Itemskr 1 400,00: 632417012600: KAVA632417012600
Specifications
Environmentally Preferable - 213-1067632417012800800 ml175 mm90 mm101 * 10 Itemskr 2 150,00: 632417012800: KAVA632417012800
Specifications
Environmentally Preferable