
2-Chloroanthracene ≥98.0% (by GC)
TCIAC1962-1G
: TCI
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:222.0 to 226.0 deg-C
:
:212.68 g/mol
:347 °C (1013 hPa)
:223 °C
:GHS07
:17135-78-3
:C₁₄H₉Cl
:MFCD00003580