Norway
Order Entry
trans-2-Dodecen-1-ol ≥90.0% (by GC)
  TCIAD1936-5ML
 :  TCI
 :  
undefined
trans-2-Dodecen-1-ol ≥90.0% (by GC)
  TCIAD1936-5ML
 :  TCI
 :  
 
kr 2 500,00
   
  1 * 5 mL

 

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

 

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 90.0 %

 

 

 :
 :184.3184 g/mol
 :141°C
 :3°C
 :69064-37-5
 :C₁₂H₂₄O
 :MFCD00014047