Solid phase extraction cartridges, Bond Elut™ Focus
: VARIAN
Bond Elut™ Focus solid phase extraction cartridges with enhanced polar recovery. Features a unique amide functionalised polar-enhanced sorbent technology that delivers outstanding retention for polar and non-polar analytes.
- Simple, universal method reduces method validation times
- 'Power Rinse' delivers cleaner extracts, improving sensitivity
- Hydrogen bond donor capability increases retention of basic analytes
Rigorous washes using 10 to 20% ACN and MeOH can be used to aggressively remove endogenous matrix interferences. For very polar analytes that are held by strictly polar interactions.
- PhasePore sizeParticle SizeCapacityBed weightPk
- Straight barrel cartridgeStraight barrel cartridgeVARIA5306021A5306021Focus120 Å44 µm3 ml20 mg1001 * 100 Itemskr 12 225,00
Accessories
Specifications
Environmentally Preferable - Straight barrel cartridgeStraight barrel cartridgeVARIA5306022A5306022Focus120 Å44 µm3 ml60 mg1001 * 100 Itemskr 13 375,00
Accessories
Specifications
Environmentally Preferable - Straight barrel cartridgeStraight barrel cartridgeVARIA5106010A5106010Focus120 Å44 µm1 ml10 mg31 * 3 Itemskr 9 850,00
Accessories
Specifications
Environmentally Preferable - Straight barrel cartridgeStraight barrel cartridgeVARIA5606050A5606050Focus120 Å44 µm6 ml50 mg1001 * 100 Itemskr 22 175,00
Accessories
Specifications
Environmentally Preferable