HPLC columns, SeQuant® ZIC®-cHILIC
: Merck
ZIC®-cHILIC is based on the same technology platform as ZIC®-HILIC, but has a different functional group and an improved manufacturing procedure. The different charge orientation of the zwitterionic functional group leads to a different – and complementary – selectivity for polar hydrophilic compounds. This selectivity is especially beneficial for hydrophilic compounds that are negatively charged, positively charged, zwitterionic or neutral, for example, organic acids, amino acids, metabolites, aminoglycosides and nucleotides.
- Complementary selectivity by reoriented charges in zwitterionic functional group
- Perfect LC-MS compatibility due to low eluent buffer strengths and very low bleeding
- High density-bonded phosphorylcholine zwitterionic stationary phase
- Very high stability and excellent batch-to-batch reproducibility
ZIC®-cHILIC, analytical columns
- Column hardwarePore sizeParticle SizeØ int.Length
- ZIC®-cHILIC HPLC columnZIC®-cHILIC HPLC column1.50656.0001Stainless steel, PEEK coated100 Å3 µm2,10 mm50 mm1 * 1 Itemskr 9 510,00: 1.50656.0001: MERC1.50656.0001
Specifications
Environmentally Preferable - ZIC®-cHILIC HPLC columnZIC®-cHILIC HPLC column1.50657.0001Stainless steel, PEEK coated100 Å3 µm2,10 mm100 mm1 * 1 Itemskr 13 580,00: 1.50657.0001: MERC1.50657.0001
Specifications
Environmentally Preferable - ZIC®-cHILIC HPLC columnZIC®-cHILIC HPLC column1.50658.0001Stainless steel, PEEK coated100 Å3 µm2,10 mm150 mm1 * 1 Itemskr 14 940,00: 1.50658.0001: MERC1.50658.0001
Specifications
Environmentally Preferable - ZIC®-cHILIC HPLC validation kit (3 columns of 3 different batches)ZIC®-cHILIC HPLC validation kit (3 columns of 3 different batches)1.50658.1003Stainless steel, PEEK coated100 Å3 µm2,10 mm150 mm1 * 1 Itemskr 40 150,00: 1.50658.1003: MERC1.50658.1003
Specifications
Environmentally Preferable - ZIC®-cHILIC HPLC columnZIC®-cHILIC HPLC column1.50659.0001Stainless steel, PEEK coated100 Å3 µm4,60 mm50 mm1 * 1 Itemskr 9 400,00: 1.50659.0001: MERC1.50659.0001
Specifications
Environmentally Preferable - ZIC®-cHILIC HPLC columnZIC®-cHILIC HPLC column1.50662.0001Stainless steel, PEEK coated100 Å3 µm4,60 mm250 mm1 * 1 Itemskr 18 900,00: 1.50662.0001: MERC1.50662.0001
Specifications
Environmentally Preferable - ZIC®-cHILIC HPLC columnZIC®-cHILIC HPLC column1.50660.0001Stainless steel, PEEK coated100 Å3 µm4,60 mm100 mm1 * 1 Itemskr 13 580,00: 1.50660.0001: MERC1.50660.0001
Specifications
Environmentally Preferable - ZIC®-cHILIC HPLC columnZIC®-cHILIC HPLC column1.50661.0001Stainless steel, PEEK coated100 Å3 µm4,60 mm150 mm1 * 1 Itemskr 17 950,00: 1.50661.0001: MERC1.50661.0001
Specifications
Environmentally Preferable
ZIC®-cHILIC, capillary columns
- Column hardwarePore sizeParticle SizeØ int.Length
- ZIC®-cHILIC capillary HPLC columnZIC®-cHILIC capillary HPLC column1.50669.0001Stainless steel, PEEK coated100 Å3 µm0,30 mm150 mm1 * 1 Itemskr 13 050,00: 1.50669.0001: MERC1.50669.0001
Specifications
Environmentally Preferable - ZIC®-cHILIC capillary HPLC columnZIC®-cHILIC capillary HPLC column1.50670.0001Stainless steel, PEEK coated100 Å3 µm1,00 mm150 mm1 * 1 Itemskr 12 175,00: 1.50670.0001: MERC1.50670.0001
Specifications
Environmentally Preferable