Norway
Order Entry
VWR®, Holmium standard solution, 1,000 mg/L Ho in dilute nitric acid AVS TITRINORM standard for AAS
VWR®, Holmium standard solution, 1,000 mg/L Ho in dilute nitric acid AVS TITRINORM standard for AAS
Supplier:  VWR Chemicals
Certificates
undefined
VWR®, Holmium standard solution, 1,000 mg/L Ho in dilute nitric acid AVS TITRINORM standard for AAS
Supplier:  VWR Chemicals
Certificates
Related Information
Product Information
  • Catalog #
    Element
    Symbol
    Concentration
    Matrix
    Pack type
    Pk
    Availability
    Price
  • Holmium
    Ho
    1000 mg/L
    Nitric acid dil.
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 985,00
    kr 985,00
    1 * 500 mL
       kr 985,00
    Local Article Number: VWRC86683.260
  • Holmium
    Ho
    1000 mg/L
    Nitric acid dil.
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - kr 605,00
    kr 605,00
    1 * 100 mL
       kr 605,00
    Local Article Number: VWRC86683.180
Frequently Bought Together