Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Norway
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
Order Entry
Home
Shoes and Boots
Safety shoes, slip-on, Bacou Pro'Clean Black, sperian
Safety shoes, slip-on, Bacou Pro'Clean Black, sperian
Safety shoes, slip-on, Bacou Pro'Clean Black, sperian
:
HONEYWELL SAFETY
Safety shoes, slip-on, Bacou Pro'Clean Black, sperian
:
HONEYWELL SAFETY
Shoe size
Colour
Pk
114-3387
6246130/44
44
Black
1 PAIR
1 * 1 PAR
-
+
1 * 1 PAR
-
+
Specifications
Environmentally Preferable
114-3382
6246130/39
39
Black
1 PAIR
1 * 1 PAR
-
+
1 * 1 PAR
-
+
Specifications
Environmentally Preferable
114-3383
6246130/40
40
Black
1 PAIR
1 * 1 PAR
-
+
1 * 1 PAR
-
+
Specifications
Environmentally Preferable