Stopcocks, high vacuum, angled
Leverandør: SciLabware
BIBBSSV504EA 1200 NOK
BIBBSSV504
Stopcocks, high vacuum, angled
Pump pattern stopcock, right angled, made of borosilicate glass for high vacuum application.
  • Connects vacuum line to a pump mounted at a lower level
  • Pump holds the key securely when the stopcock is closed
  • Side-arm length of 100 mm