Flash cartridges, Mega Bond Elut™
: VARIAN
Flash cartridges for the purification of organic compounds, and also for scale-up, solid phase extraction.
- Convenient disposable cartridges eliminate the need for packing glass columns
- Flexible 'open' tube design for either liquid or solid samples
- Reliable, consistent flow characteristics deliver high-resolution performance
- Column FormatPhaseParticle SizeCapacityBed weightPk
- 551-040912256015Flash cartridgeC1840 µm12 ml2 g201 * 20 Itemskr 5 500,00: 12256015: VARI12256015
Specifications
Environmentally Preferable - 551-041012256023Flash cartridgeC1840 µm20 ml5 g201 * 20 Itemskr 9 875,00: 12256023: VARI12256023
Specifications
Environmentally Preferable - 551-041512256031Flash cartridgeC1840 µm60 ml10 g161 * 16 Itemskr 12 775,00: 12256031: VARI12256031
Specifications
Environmentally Preferable - 557-001112256078Flash cartridgeC1840 µm60 ml20 g161 * 16 Itemskr 26 500,00: 12256078: VARI12256078
Specifications
Environmentally Preferable - 557-001212256079Flash cartridgeC1840 µm150 ml25 g81 * 8 Itemskr 15 450,00: 12256079: VARI12256079
Specifications
Environmentally Preferable - 557-001312256080Flash cartridgeC1840 µm150 ml50 g81 * 8 Itemskr 30 325,00: 12256080: VARI12256080
Specifications
Environmentally Preferable - 557-001412256081Flash cartridgeC1840 µm150 ml70 g81 * 8 Itemskr 35 800,00: 12256081: VARI12256081
Specifications
Environmentally Preferable - VARI1225602012256020Flash cartridgeNH₂40 µm12 ml2 g201 * 20 Itemskr 6 700,00: 12256020: VARI12256020
Specifications
Environmentally Preferable - 551-041312256028Flash cartridgeNH₂40 µm20 ml5 g201 * 20 Itemskr 10 950,00: 12256028: VARI12256028
Specifications
Environmentally Preferable - 557-000712256074Flash cartridgeNH₂40 µm60 ml20 g161 * 16 Itemskr 26 000,00: 12256074: VARI12256074
Specifications
Environmentally Preferable - 557-000812256075Flash cartridgeNH₂40 µm150 ml25 g81 * 8 Itemskr 15 300,00: 12256075: VARI12256075
Specifications
Environmentally Preferable - 557-000912256076Flash cartridgeNH₂40 µm150 ml50 g81 * 8 Itemskr 30 325,00: 12256076: VARI12256076
Specifications
Environmentally Preferable - 557-001012256077Flash cartridgeNH₂40 µm150 ml70 g81 * 8 Itemskr 42 450,00: 12256077: VARI12256077
Specifications
Environmentally Preferable - 557-000312256070Flash cartridgeSCX40 µm150 ml25 g81 * 8 Itemskr 15 300,00: 12256070: VARI12256070
Specifications
Environmentally Preferable - 557-000512256072Flash cartridgeSCX40 µm150 ml50 g81 * 8 Itemskr 30 325,00: 12256072: VARI12256072
Specifications
Environmentally Preferable - 557-000612256073Flash cartridgeSCX40 µm150 ml70 g81 * 8 Itemskr 42 450,00: 12256073: VARI12256073
Specifications
Environmentally Preferable - VARI1225601812256018Flash cartridgeSI40 µm12 ml2 g201 * 20 Itemskr 4 450,00: 12256018: VARI12256018
Specifications
Environmentally Preferable - 551-041212256026Flash cartridgeSI40 µm20 ml5 g201 * 20 Itemskr 5 725,00: 12256026: VARI12256026
Specifications
Environmentally Preferable - VARI1225603412256034Flash cartridgeSI40 µm60 ml10 g161 * 16 Itemskr 6 300,00: 12256034: VARI12256034
Specifications
Environmentally Preferable - VARI1225604212256042Flash cartridgeSI40 µm60 ml20 g161 * 16 Itemskr 7 975,00: 12256042: VARI12256042
Specifications
Environmentally Preferable - 557-000112256067Flash cartridgeSI40 µm150 ml50 g81 * 8 Itemskr 8 300,00: 12256067: VARI12256067
Specifications
Environmentally Preferable - VARI1225606812256068Flash cartridgeSI40 µm60 ml15 g161 * 16 Itemskr 6 875,00: 12256068: VARI12256068
Specifications
Environmentally Preferable - 557-000212256069Flash cartridgeSI40 µm150 ml25 g81 * 8 Itemskr 5 550,00: 12256069: VARI12256069
Specifications
Environmentally Preferable - 557-000412256071Flash cartridgeSI40 µm150 ml70 g81 * 8 Itemskr 10 025,00: 12256071: VARI12256071
Specifications
Environmentally Preferable